Thermo Scientific Chemicals

2-Bromo-4,6-difluoroaniline, 98%, Thermo Scientific Chemicals

Catalog number: A11022.14
25 g, Each
Thermo Scientific Chemicals

2-Bromo-4,6-difluoroaniline, 98%, Thermo Scientific Chemicals

Catalog number: A11022.14
25 g, Each
Quantity
Catalog number: A11022.14
also known as A11022-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
444-14-4
IUPAC Name
2-bromo-4,6-difluoroaniline
Molecular Formula
C6H4BrF2N
InChI Key
WUJKFVGKLTWVSQ-UHFFFAOYSA-N
SMILES
NC1=C(F)C=C(F)C=C1Br
Appearance (Color)
White to pale brown or grey to purple
Form
crystals or crystalline powder
Assay (GC)
>97.5%

Description

2-Bromo-4,6-difluoroaniline is used in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4,6-difluoroaniline is used in chemical synthesis.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text