4-Chlorothioanisole, 98%, Thermo Scientific Chemicals
4-Chlorothioanisole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chlorothioanisole, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A11025.14
also known as A11025-14
Price (USD)/ Each
147.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
147.00
Add to cart
4-Chlorothioanisole, 98%, Thermo Scientific Chemicals
Catalog numberA11025.14
Price (USD)/ Each
147.00
-
Add to cart
Chemical Identifiers
CAS123-09-1
IUPAC Name1-chloro-4-(methylsulfanyl)benzene
Molecular FormulaC7H7ClS
InChI KeyKIQQUVJOLVCZKG-UHFFFAOYSA-N
SMILESCSC1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Clear colorless to pale yellow
CommentMay solidify in cold temperatures, but will melt if allowed to warm to room temperature.
Refractive Index1.5980-1.6020 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only