Thermo Scientific Chemicals

Cyclopentene, 97%, Thermo Scientific Chemicals

Catalog number: A11027.AP
500 mL, Each
Thermo Scientific Chemicals

Cyclopentene, 97%, Thermo Scientific Chemicals

Catalog number: A11027.AP
500 mL, Each
Quantity
Catalog number: A11027.AP
also known as A11027-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
142-29-0
IUPAC Name
cyclopentene
Molecular Formula
C5H8
InChI Key
LPIQUOYDBNQMRZ-UHFFFAOYSA-N
SMILES
C1CC=CC1
Refractive Index
1.4195-1.4245 @ 20?C
Form
Liquid
Appearance (Color)
Clear colorless
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms

Description

Cyclopentene is used in the production of cyclopentene oxide, ketamine, cyclopentyl phenol (disinfectant), aldehyde and cyclopentene glycol. It acts as a monomer and is utilized for the synthesis of plastics and in various chemical syntheses. It is also used in the investigation of [2+2] cycloaddition of diamond (001) surfaces with alkene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentene is used in the production of cyclopentene oxide, ketamine, cyclopentyl phenol (disinfectant), aldehyde and cyclopentene glycol. It acts as a monomer and is utilized for the synthesis of plastics and in various chemical syntheses. It is also used in the investigation of [2+2] cycloaddition of diamond (001) surfaces with alkene.

Solubility
Miscible with ethanol, ether, benzene and organic solvents. Immiscible with water.

Notes
Store in cool place. Air sensitive. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text