Tryptamine hydrochloride, 98+%, Thermo Scientific Chemicals
Tryptamine hydrochloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tryptamine hydrochloride, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A11030.18
also known as A11030-18
Price (USD)/ Each
226.65
Online exclusive
266.00 
Save 39.35 (15%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
226.65
Online exclusive
266.00 
Save 39.35 (15%)
Add to cart
Tryptamine hydrochloride, 98+%, Thermo Scientific Chemicals
Catalog numberA11030.18
Price (USD)/ Each
226.65
Online exclusive
266.00 
Save 39.35 (15%)
-
Add to cart
Chemical Identifiers
CAS343-94-2
IUPAC Namehydrogen 2-(1H-indol-3-yl)ethan-1-amine chloride
Molecular FormulaC10H13ClN2
InChI KeyKDFBGNBTTMPNIG-UHFFFAOYSA-N
SMILES[H+].[Cl-].NCCC1=CNC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to pale orange
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥98.0 to ≤102.0%
Melting Point (clear melt)250.0-257.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only