Thermo Scientific Chemicals

Diethylenetriamine, 99%, Thermo Scientific Chemicals

Catalog number: A11033.AP
500 mL, Each
Thermo Scientific Chemicals

Diethylenetriamine, 99%, Thermo Scientific Chemicals

Catalog number: A11033.AP
500 mL, Each
Quantity
Catalog number: A11033.AP
also known as A11033-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-40-0
IUPAC Name
bis(2-aminoethyl)amine
Molecular Formula
C4H13N3
InChI Key
RPNUMPOLZDHAAY-UHFFFAOYSA-N
SMILES
NCCNCCN
Appearance (Color)
Clear colorless to pale yellow or pale pink or pale brown
Assay (GC)
≥98.5% (UK-sourced material)
Assay from Supplier's CofA
≥98.5% (US-sourced Material)
Comment
Material sourced in UK and US
Identification (FTIR)
Conforms (UK-sourced material)

Description

Diethylenetriamine is used as a solvent for plastics, dyes and in chemical synthesis. It serves as a curing agent for epoxy-resins. As a tridentate ligand, it form complexes such as Co(dien)(NO2)3 in coordination chemistry. It is also used as a fuel additive. It reacts with fatty acid resulting in the formation of amidoamines, which are used as corrosion inhibitors in the petroleum industry. It is associated with unsymmetrical dimethylhydrazine to prepare hydyne, which is a propellant for liquid-fuel rockets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylenetriamine is used as a solvent for plastics, dyes and in chemical synthesis. It serves as a curing agent for epoxy-resins. As a tridentate ligand, it form complexes such as Co(dien)(NO2)3 in coordination chemistry. It is also used as a fuel additive. It reacts with fatty acid resulting in the formation of amidoamines, which are used as corrosion inhibitors in the petroleum industry. It is associated with unsymmetrical dimethylhydrazine to prepare hydyne, which is a propellant for liquid-fuel rockets.

Solubility
Miscible with water and ethanol. Immiscible with ether.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids and copper.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text