Thermo Scientific Chemicals

Dimethyl cyanodithioiminocarbonate, 95%, Thermo Scientific Chemicals

Catalog number: A11043.14
25 g, Each
Thermo Scientific Chemicals

Dimethyl cyanodithioiminocarbonate, 95%, Thermo Scientific Chemicals

Catalog number: A11043.14
25 g, Each
Quantity
Catalog number: A11043.14
also known as A11043-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10191-60-3
IUPAC Name
[bis(methylsulfanyl)methylidene](cyano)amine
Molecular Formula
C4H6N2S2
InChI Key
IULFXBLVJIPESI-UHFFFAOYSA-N
SMILES
CSC(SC)=NC#N
Appearance (Color)
White to yellow or pale brown
Form
Powder and/or Lumps
Assay (GC)
>94.0%
Water Content (Karl Fischer Titration)
<2.0%

Description

Dimethyl cyanodithioiminocarbonate has been used in the synthesis of 4-methylthiopyrazolo[1,5-a]-1,3,5-triazines, methylsulfanylpyrimidines, cyanoguanidines and N-aryl-6-methylsulfanyl-4-oxopyrimidine-5-carbonitriles. It has been also used in the synthesis of methylsulfanyl derivatives of azoloazines and azoloazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl cyanodithioiminocarbonate has been used in the synthesis of 4-methylthiopyrazolo[1,5-a]-1,3,5-triazines, methylsulfanylpyrimidines, cyanoguanidines and N-aryl-6-methylsulfanyl-4-oxopyrimidine-5-carbonitriles. It has been also used in the synthesis of methylsulfanyl derivatives of azoloazines and azoloazoles.

Solubility
Slightly soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text