Thermo Scientific Chemicals

S-Methylisothiouronium sulfate, 98+%, Thermo Scientific Chemicals

Catalog number: A11044.36
500 g, Each
Thermo Scientific Chemicals

S-Methylisothiouronium sulfate, 98+%, Thermo Scientific Chemicals

Catalog number: A11044.36
500 g, Each
Quantity
Catalog number: A11044.36
also known as A11044-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
867-44-7
IUPAC Name
bis((methylsulfanyl)methanimidamide); sulfuric acid
Molecular Formula
C4H14N4O4S3
InChI Key
BZZXQZOBAUXLHZ-UHFFFAOYSA-N
SMILES
CSC(N)=N.CSC(N)=N.OS(O)(=O)=O
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0%
Form
Crystals or powder or crystalline powder or fused solid
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text