Thermo Scientific Chemicals

Cyanoacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: A11045.0B
1000 g, Each
Thermo Scientific Chemicals

Cyanoacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: A11045.0B
1000 g, Each
Quantity
Catalog number: A11045.0B
also known as A11045-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
372-09-8
IUPAC Name
2-cyanoacetic acid
Molecular Formula
C3H3NO2
InChI Key
MLIREBYILWEBDM-UHFFFAOYSA-N
SMILES
OC(=O)CC#N
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Form
Crystals or powder or crystalline powder or fused/lumpy solid
Appearance (Color)
White to cream to pale brown

Description

Cyanoacetic acid is used in pharmaceutical industry for producing vitamin B6 and caffeine. It is also used in making dyes, agricultural chemicals, and in the synthesis of phenylacetic acid esters. It acts as a precursor cyanoacrylates viz. ethyl cyanoacrylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyanoacetic acid is used in pharmaceutical industry for producing vitamin B6 and caffeine. It is also used in making dyes, agricultural chemicals, and in the synthesis of phenylacetic acid esters. It acts as a precursor cyanoacrylates viz. ethyl cyanoacrylate.

Solubility
Miscible with water, ethyl alcohol, ether. Slightly miscible with benzene, acetic acid and chloroform.

Notes
Hygroscopic. Incompatible with strong bases, acids, strong oxidizing agents, alkaline, nitriles and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text