Thermo Scientific Chemicals

4-Amidinopyridine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: A11050.06
5 g, Each
Thermo Scientific Chemicals

4-Amidinopyridine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: A11050.06
5 g, Each
Quantity
Catalog number: A11050.06
also known as A11050-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6345-27-3
IUPAC Name
pyridine-4-carboximidamide hydrochloride
Molecular Formula
C6H8ClN3
InChI Key
IONKMFGAXKCLMI-UHFFFAOYSA-N
SMILES
Cl.NC(=N)C1=CC=NC=C1
Form
Crystals or powder or crystalline powder
Assay from Supplier's CofA
≥98.0% (HPLC) (U.S. specification)
Water Content (Karl Fischer Titration)
≤1% (U.K. specification)
Appearance (Color)
White
Assay (Titration ex Chloride)
≥98.0 to ≤102.0% (U.K. specification)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text