2-Nitroaniline is the main precursor to phenylenediamines, which are converted to benzimidazoles, a family of heterocycles that are key components in pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Nitroaniline is the main precursor to phenylenediamines, which are converted to benzimidazoles, a family of heterocycles that are key components in pharmaceuticals.
Solubility
Soluble in water (0.6 g/L at 20°C).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Kunquan Lia; Zheng Zheng; Xingfa Huang; Guohua Zhao; Jingwei Feng; Jibiao Zhang. Equilibrium, kinetic and thermodynamic studies on the adsorption of 2-nitroaniline onto activated carbon prepared from cotton stalk fibre. Journal of Hazardous Materials. 2009, 166,(1), 213-220.
Yiming Zhang; Xun Yuan; Yuan Wang; Yi Chen. One-pot photochemical synthesis of graphene composites uniformly deposited with silver nanoparticles and their high catalytic activity towards the reduction of 2-nitroaniline. J. Mater. Chem. 2012, 22,(15), 7245-7251.