Thermo Scientific Chemicals

1-Indanone, 99+%, Thermo Scientific Chemicals

Catalog number: A11056.14
25 g, Each
Thermo Scientific Chemicals

1-Indanone, 99+%, Thermo Scientific Chemicals

Catalog number: A11056.14
25 g, Each
Quantity
Catalog number: A11056.14
also known as A11056-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
83-33-0
IUPAC Name
2,3-dihydro-1H-inden-1-one
Molecular Formula
C9H8O
InChI Key
QNXSIUBBGPHDDE-UHFFFAOYSA-N
SMILES
O=C1CCC2=CC=CC=C12
Appearance (Color)
White to pale yellow or pale brown
Assay (GC)
≥99.0%
Form
Crystals or powder or crystalline powder or fused solid or chunks or lumps
Solution Test
2.5% w/v solution in ethanol, 99.5% will be transparent to very faintly turbid by JIS K 8001 5.2

Description

1-Indanone is an oxidation product of indan, a component of fuels, solvents and varnishes. 1-Indanone is also a metabolite of Thalidomide that has been shown to inhibit the attachment of tumor cells to concanavalin A coated plastic surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Indanone is an oxidation product of indan, a component of fuels, solvents and varnishes. 1-Indanone is also a metabolite of Thalidomide that has been shown to inhibit the attachment of tumor cells to concanavalin A coated plastic surfaces.

Solubility
Soluble in water (6.5 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text