Thermo Scientific Chemicals

1-Fluoro-4-nitrobenzene, 99%, Thermo Scientific Chemicals

Catalog number: A11057.36
500 g, Each
Thermo Scientific Chemicals

1-Fluoro-4-nitrobenzene, 99%, Thermo Scientific Chemicals

Catalog number: A11057.36
500 g, Each
Quantity
Catalog number: A11057.36
also known as A11057-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
350-46-9
IUPAC Name
1-fluoro-4-nitrobenzene
Molecular Formula
C6H4FNO2
InChI Key
WFQDTOYDVUWQMS-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(F)C=C1
Assay (GC)
> 98.5%
Form
liquid as melt
Clarity
25% in ethanol, clear solution
Refractive Index
1.5300 - 1.5330 @20?C
Appearance (Color)
Clear, yellow

Description

1-Fluoro-4-nitrobenzene used as a component of hair dyes, as pharmaceutical intermediates. It is also used for the preparation of novel soluble aromatic polyimides

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Fluoro-4-nitrobenzene used as a component of hair dyes, as pharmaceutical intermediates. It is also used for the preparation of novel soluble aromatic polyimides

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents, acids, acid chlorides, acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text