Thermo Scientific Chemicals

Tri-n-octylphosphine oxide, 98%, Thermo Scientific Chemicals

Catalog number: A11062.36
500 g, Each
Thermo Scientific Chemicals

Tri-n-octylphosphine oxide, 98%, Thermo Scientific Chemicals

Catalog number: A11062.36
500 g, Each
Quantity
Catalog number: A11062.36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78-50-2
IUPAC Name
1-(dioctylphosphoryl)octane
Molecular Formula
C24H51OP
InChI Key
ZMBHCYHQLYEYDV-UHFFFAOYSA-N
SMILES
CCCCCCCCP(=O)(CCCCCCCC)CCCCCCCC
Melting Point (clear melt)
50-57?C
Identification (FTIR)
Conforms
Appearance (Color)
White to cream or pale yellow
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder or flakes and/or fused solid/chunks

Description

Tri-n-octylphosphine oxide is a solvent used in the extraction of metals, especially uranium, zirconium and hafnium. It is also used as an auxiliary reagent in the solvent extraction of metals. It is also used to extract hydrogen bonding organic compounds. Further, it is useful as a capping ligand for the production of quantum dots, such as those consisting of cadmium selenide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-octylphosphine oxide is a solvent used in the extraction of metals, especially uranium, zirconium and hafnium. It is also used as an auxiliary reagent in the solvent extraction of metals. It is also used to extract hydrogen bonding organic compounds. Further, it is useful as a capping ligand for the production of quantum dots, such as those consisting of cadmium selenide.

Solubility
Soluble in chloroform, toluene and hexane. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text