Thermo Scientific Chemicals

1-Naphthylacetic acid, 95%, may cont. up to 5% 2-isomer, Thermo Scientific Chemicals

Catalog number: A11069.0B
1000 g, Each
Thermo Scientific Chemicals

1-Naphthylacetic acid, 95%, may cont. up to 5% 2-isomer, Thermo Scientific Chemicals

Catalog number: A11069.0B
1000 g, Each
Quantity
Catalog number: A11069.0B
also known as A11069-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
86-87-3
Appearance (Color)
White to cream to yellow or pale brown
Form
Crystalline powder or powder
Assay (Aqueous acid-base Titration)
≥94.0 to ≤106.0% (sum of 1-isomer & 2-isomer)
Assay (Silylated GC)
≥94.0% (1-isomer)
Identification (FTIR)
Conforms

Description

1-Naphthylacetic acid is used as a plant growth regulator and for plant tissue culture. It is used for the vegetative propagation of plants from stem and leaf cutting, abscission promoter and substrate for auxin inducers of specific plant enzymes. Further, it stimulates root formation in cuttings of woody and herbaceous plants. It is also used to prevent premature flower and fruit drop. It is also employed to control regrowth of tree sprouts after trimminguse. In addition to this, it is used in ornaments and serves as an ingredient in many commercial plant rooting horticultural products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthylacetic acid is used as a plant growth regulator and for plant tissue culture. It is used for the vegetative propagation of plants from stem and leaf cutting, abscission promoter and substrate for auxin inducers of specific plant enzymes. Further, it stimulates root formation in cuttings of woody and herbaceous plants. It is also used to prevent premature flower and fruit drop. It is also employed to control regrowth of tree sprouts after trimminguse. In addition to this, it is used in ornaments and serves as an ingredient in many commercial plant rooting horticultural products.

Solubility
Slightly soluble in water, ethanol, acetone, chloroform and ether.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text