Thermo Scientific™

Xanthine, 99%, Thermo Scientific Chemicals

Catalog number: A11077.22
100 g, Each
Thermo Scientific™

Xanthine, 99%, Thermo Scientific Chemicals

Catalog number: A11077.22
100 g, Each
Quantity
Catalog number: A11077.22
also known as A11077-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
69-89-6
IUPAC Name
2,3,6,7-tetrahydro-1H-purine-2,6-dione
Molecular Formula
C5H4N4O2
InChI Key
LRFVTYWOQMYALW-UHFFFAOYSA-N
SMILES
O=C1NC2=C(NC=N2)C(=O)N1
Appearance (Color)
White to pale cream
Assay from Supplier's CofA
≥98.5% (HPLC)
Form
Powder
Infrared spectrum
Conforms

Description

Xanthine and xanthine oxidase system can be used to produce superoxide radicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Xanthine and xanthine oxidase system can be used to produce superoxide radicals.

Solubility
Soluble in water(0.067g/L).

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text