3-Acetylthiophene, 98%, Thermo Scientific Chemicals
3-Acetylthiophene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Acetylthiophene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Catalog number A11078.18
also known as A11078-18
Price (USD)/ Each
368.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
368.00
Add to cart
3-Acetylthiophene, 98%, Thermo Scientific Chemicals
Catalog numberA11078.18
Price (USD)/ Each
368.00
-
Add to cart
Chemical Identifiers
CAS1468-83-3
IUPAC Name1-(thiophen-3-yl)ethan-1-one
Molecular FormulaC6H6OS
InChI KeyRNIDWJDZNNVFDY-UHFFFAOYSA-N
SMILESCC(=O)C1=CSC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (GC)≥97.5%
Melting Point (clear melt)57.0-63.0?C
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only