Salicylaldoxime, 98%, Thermo Scientific Chemicals
Salicylaldoxime, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Salicylaldoxime, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A11080.14
also known as A11080-14
Price (USD)/ Each
35.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
35.00
Add to cart
Salicylaldoxime, 98%, Thermo Scientific Chemicals
Catalog numberA11080.14
Price (USD)/ Each
35.00
-
Add to cart
Chemical Identifiers
CAS94-67-7
IUPAC Name(6Z)-6-[(hydroxyamino)methylidene]cyclohexa-2,4-dien-1-one
Molecular FormulaC7H7NO2
InChI KeyGFCNBJDXQMZOOC-WAYWQWQTSA-N
SMILESON\C=C1\C=CC=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White to pale cream to cream to pale brown
FormCrystals or powder or crystalline powder
Melting Point (clear melt)54.0-60.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only