Thermo Scientific Chemicals

Salicylaldoxime, 98%, Thermo Scientific Chemicals

Catalog number: A11080.36
500 g, Each
Thermo Scientific Chemicals

Salicylaldoxime, 98%, Thermo Scientific Chemicals

Catalog number: A11080.36
500 g, Each
Quantity
Catalog number: A11080.36
also known as A11080-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
94-67-7
IUPAC Name
(6Z)-6-[(hydroxyamino)methylidene]cyclohexa-2,4-dien-1-one
Molecular Formula
C7H7NO2
InChI Key
GFCNBJDXQMZOOC-WAYWQWQTSA-N
SMILES
ON\C=C1\C=CC=CC1=O
Assay (GC)
≥97.5%
Appearance (Color)
White to pale cream to cream to pale brown
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
54.0-60.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text