Thermo Scientific Chemicals

D-(+)-Glucose monohydrate

Catalog number: A11090.0E
2500 g, Each
Thermo Scientific Chemicals

D-(+)-Glucose monohydrate

Catalog number: A11090.0E
2500 g, Each
Quantity
Catalog number: A11090.0E
also known as A11090-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14431-43-7
IUPAC Name
(2S,3R,4S,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol hydrate
Molecular Formula
C6H14O7
InChI Key
OSNSWKAZFASRNG-WNFIKIDCSA-N
SMILES
O.OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O
Identification (FTIR)
Conforms
Appearance (Color)
White
Water Content (Karl Fischer Titration)
≥8.0 to ≤9.6%
Form
Crystals or powder or crystalline powder
Optical Rotation
+51° to +54° (c=10 in water, dry wt. basis)

Description

D-(+)-Glucose monohydrate is used by industry professionals to indicate the degree to which a substance contains water, and in food industry, pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(+)-Glucose monohydrate is used by industry professionals to indicate the degree to which a substance contains water, and in food industry, pharmaceuticals.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text