Thermo Scientific Chemicals

Tris(2-methoxyphenyl)phosphine, 98%, Thermo Scientific Chemicals

Catalog number: A11096.06
5 g, Each
Thermo Scientific Chemicals

Tris(2-methoxyphenyl)phosphine, 98%, Thermo Scientific Chemicals

Catalog number: A11096.06
5 g, Each
Quantity
Catalog number: A11096.06
also known as A11096-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4731-65-1
IUPAC Name
tris(2-methoxyphenyl)phosphane
Molecular Formula
C21H21O3P
InChI Key
IIOSDXGZLBPOHD-UHFFFAOYSA-N
SMILES
COC1=CC=CC=C1P(C1=CC=CC=C1OC)C1=CC=CC=C1OC
Form
Powder
Appearance (Color)
White
Assay (GC)
≥97.5%
Melting Point (clear melt)
190-200°C

Description

Tris(2-methoxyphenyl)phosphine is a phosphine ligand used for catalysis. It is used as a catalyst for allylic substitution of simple alkenes, suzuki coupling reactions, hydrogenation of quinolines and other addition reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(2-methoxyphenyl)phosphine is a phosphine ligand used for catalysis. It is used as a catalyst for allylic substitution of simple alkenes, suzuki coupling reactions, hydrogenation of quinolines and other addition reactions.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text