Thermo Scientific Chemicals

1,4-Cyclohexanedione, 98%, Thermo Scientific Chemicals

Catalog number: A11100.14
25 g, Each
Thermo Scientific Chemicals

1,4-Cyclohexanedione, 98%, Thermo Scientific Chemicals

Catalog number: A11100.14
25 g, Each
Quantity
Catalog number: A11100.14
also known as A11100-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
637-88-7
IUPAC Name
cyclohexane-1,4-dione
Molecular Formula
C6H8O2
InChI Key
DCZFGQYXRKMVFG-UHFFFAOYSA-N
SMILES
O=C1CCC(=O)CC1
Melting Point (clear melt)
74.5-80.5?C
Form
Crystals or powder or crystalline powder
Appearance (Color)
Pale yellow to yellow or pale cream to pale brown
Assay (GC)
≥97.5%

Description

1,4-Cyclohexanedione is used in the preparation of 1,4 benzoquinone and bromoorganics. It is also used to study the influence of visible light on the bromate-1,4-cyclohexanedione-ferroin oscillating reaction. It plays a vital role in pharmaceuticals, plant growth regulator and as a conducting material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Cyclohexanedione is used in the preparation of 1,4 benzoquinone and bromoorganics. It is also used to study the influence of visible light on the bromate-1,4-cyclohexanedione-ferroin oscillating reaction. It plays a vital role in pharmaceuticals, plant growth regulator and as a conducting material.

Solubility
Soluble in methanol and ethanol.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text