Thermo Scientific Chemicals

4-Vinylbenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A11107.06
5 g, Each
Thermo Scientific Chemicals

4-Vinylbenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A11107.06
5 g, Each
Quantity
Catalog number: A11107.06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1075-49-6
IUPAC Name
4-ethenylbenzoic acid
Molecular Formula
C9H8O2
InChI Key
IRQWEODKXLDORP-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(C=C)C=C1
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (Silylated GC)
≥97.5%
Form
Crystals or powder or crystalline powder
Solution Test
(10% w/v in methanol) : Clear
Appearance (Color)
White to cream to brown

Description

It is used as a metabolite of 1,4-diethenylbenzene. It is also an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a metabolite of 1,4-diethenylbenzene. It is also an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Soluble in dichloromethane and methanol

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text