4-Vinylbenzoic acid, 98%, Thermo Scientific Chemicals
4-Vinylbenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Vinylbenzoic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number A11107.14
also known as A11107-14
Price (USD)/ Each
1,284.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
1,284.00
Add to cart
4-Vinylbenzoic acid, 98%, Thermo Scientific Chemicals
Catalog numberA11107.14
Price (USD)/ Each
1,284.00
-
Add to cart
Chemical Identifiers
CAS1075-49-6
IUPAC Name4-ethenylbenzoic acid
Molecular FormulaC9H8O2
InChI KeyIRQWEODKXLDORP-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C=C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
FormCrystals or powder or crystalline powder
Solution Test(10% w/v in methanol) : Clear
Appearance (Color)White to cream to brown
It is used as a metabolite of 1,4-diethenylbenzene. It is also an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a metabolite of 1,4-diethenylbenzene. It is also an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Soluble in dichloromethane and methanol

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

General References:

  1. Padma L. Nayak,; Ke Yang,; Pradeep K. Dhal,; Shridhara Alva,; Jayant Kumar,; Sukant K. Tripathy. Polyelectrolyte-Containing Fullerene I: Synthesis and Characterization of the Copolymers of 4-Vinylbenzoic Acid with C60. Chem. Mater. 1998, 10 (8), 2058-2066.
  2. Katja Nilles,; Patrick Theato. Synthesis and polymerization of active ester monomers based on 4-vinylbenzoic acid. European Polymer Journal. 2007n , 43(7),2901-2912.