Veratraldehyde, 99%, Thermo Scientific Chemicals
Veratraldehyde, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Veratraldehyde, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
100 g
500 g
Catalog number A11110.0E
also known as A11110-0E
Price (USD)/ Each
333.65
Online exclusive
371.00 
Save 37.35 (10%)
-
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)/ Each
333.65
Online exclusive
371.00 
Save 37.35 (10%)
Add to cart
Veratraldehyde, 99%, Thermo Scientific Chemicals
Catalog numberA11110.0E
Price (USD)/ Each
333.65
Online exclusive
371.00 
Save 37.35 (10%)
-
Add to cart
Chemical Identifiers
CAS120-14-9
IUPAC Name3,4-dimethoxybenzaldehyde
Molecular FormulaC9H10O3
InChI KeyWJUFSDZVCOTFON-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=O)C=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)41.0-48.0?C
FormFlakes
Appearance (Color)White to cream to pale yellow
Assay (GC)≥98.5%
Free acid (titration)≤0.5%
Veratraldehyde was used in the preparation of 4-chloromethyl-2-(dimethoxyphenyl)-1,3-dioxolane. It was used in the synthesis of (+)-lithospermic acid, having anti-HIV activity. It forms 1:1 inclusion complexes with cyclodextrins. It reacts with 3-acetyl-2,5-dimethythiophene to yield chalcone dye, (2E)-3-(3,4-Dimethoxyphenyl)-1-(2,5-dimethylthiophen-3-yl)prop-2-en-1-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Veratraldehyde was used in the preparation of 4-chloromethyl-2-(dimethoxyphenyl)-1,3-dioxolane. It was used in the synthesis of (+)-lithospermic acid, having anti-HIV activity. It forms 1:1 inclusion complexes with cyclodextrins. It reacts with 3-acetyl-2,5-dimethythiophene to yield chalcone dye, (2E)-3-(3,4-Dimethoxyphenyl)-1-(2,5-dimethylthiophen-3-yl)prop-2-en-1-one.

Solubility
Soluble in water. Freely soluble in alcohol and ether.

Notes
Air sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Tirumala G.Varadaraju; Jih Ru Hwu. Synthesis of anti-HIV lithospermic acid by two diverse strategies. Organic & Biomolecular Chemistry. 2012, 10,(28), 5456-5465.
  2. Abdullah M.Asiri; Hadi M.Marwani; Salman A.Khan; Samy A.El-Daly. Investigation of spectroscopic behaviors of newly synthesized (2E)-3-(3,4-dimethoxyphenyl)-1-(2,5-dimethylthiophen-3-yl)prop-2-en-1-one (DDTP) dye. Journal of Fluorescence. 2013, 23,(6), 1271-1278.
  3. Can be used to protect diols as their cyclic 3,4-dimethoxybenzylidene acetals: Tetrahedron, 43, 2549 (1987).