Thermo Scientific Chemicals

Veratraldehyde, 99%, Thermo Scientific Chemicals

Catalog number: A11110.22
100 g, Each
Thermo Scientific Chemicals

Veratraldehyde, 99%, Thermo Scientific Chemicals

Catalog number: A11110.22
100 g, Each
Quantity
Catalog number: A11110.22
also known as A11110-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
120-14-9
IUPAC Name
3,4-dimethoxybenzaldehyde
Molecular Formula
C9H10O3
InChI Key
WJUFSDZVCOTFON-UHFFFAOYSA-N
SMILES
COC1=CC=C(C=O)C=C1OC
Melting Point (clear melt)
41.0-48.0?C
Form
Flakes
Appearance (Color)
White to cream to pale yellow
Assay (GC)
≥98.5%
Free acid (titration)
≤0.5%

Description

Veratraldehyde was used in the preparation of 4-chloromethyl-2-(dimethoxyphenyl)-1,3-dioxolane. It was used in the synthesis of (+)-lithospermic acid, having anti-HIV activity. It forms 1:1 inclusion complexes with cyclodextrins. It reacts with 3-acetyl-2,5-dimethythiophene to yield chalcone dye, (2E)-3-(3,4-Dimethoxyphenyl)-1-(2,5-dimethylthiophen-3-yl)prop-2-en-1-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Veratraldehyde was used in the preparation of 4-chloromethyl-2-(dimethoxyphenyl)-1,3-dioxolane. It was used in the synthesis of (+)-lithospermic acid, having anti-HIV activity. It forms 1:1 inclusion complexes with cyclodextrins. It reacts with 3-acetyl-2,5-dimethythiophene to yield chalcone dye, (2E)-3-(3,4-Dimethoxyphenyl)-1-(2,5-dimethylthiophen-3-yl)prop-2-en-1-one.

Solubility
Soluble in water. Freely soluble in alcohol and ether.

Notes
Air sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text