Thermo Scientific Chemicals

Ethyl formate, 97%, Thermo Scientific Chemicals

Catalog number: A11113.0F
2500 mL, Each
Thermo Scientific Chemicals

Ethyl formate, 97%, Thermo Scientific Chemicals

Catalog number: A11113.0F
2500 mL, Each
Quantity
Catalog number: A11113.0F
also known as A11113-0F
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-94-4
IUPAC Name
ethyl formate
Molecular Formula
C3H6O2
InChI Key
WBJINCZRORDGAQ-UHFFFAOYSA-N
SMILES
CCOC=O
Comment
Specification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)
Conforms (non-U.S. specification)
Refractive Index
1.3580-1.3630 @ 20°C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid

Description

Ethyl formate is used as a solvent for cellulose nitrate, cellulose acetate, greases and oils. It is also used as a flavor for lemonade and essences. It is involved in the fumigation of dried fruits, as an alternative to methyl bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl formate is used as a solvent for cellulose nitrate, cellulose acetate, greases and oils. It is also used as a flavor for lemonade and essences. It is involved in the fumigation of dried fruits, as an alternative to methyl bromide.

Solubility
Miscible with water, benzene, alcohol, ether, acetone and most organic solvents.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with bases and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text