Thermo Scientific Chemicals

Diphenyl sulfide, 98%, Thermo Scientific Chemicals

Catalog number: A11114.22
100 g, Each
Thermo Scientific Chemicals

Diphenyl sulfide, 98%, Thermo Scientific Chemicals

Catalog number: A11114.22
100 g, Each
Quantity
Catalog number: A11114.22
also known as A11114-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
139-66-2
IUPAC Name
(phenylsulfanyl)benzene
Molecular Formula
C12H10S
InChI Key
LTYMSROWYAPPGB-UHFFFAOYSA-N
SMILES
S(C1=CC=CC=C1)C1=CC=CC=C1
Form
Liquid
Refractive Index
1.6310-1.6350 @ 20?C
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%

Description

Diphenyl sulfide is used as a raw material for triaryl sulfonium photoinitiator, intermediates for agrichemicals, catalyst, metal extractants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl sulfide is used as a raw material for triaryl sulfonium photoinitiator, intermediates for agrichemicals, catalyst, metal extractants.

Solubility
Soluble in ether, benzene, carbon disulphide.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents, acids, acid chlorides, acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text