Thermo Scientific Chemicals

Antimony(III) oxide, 99%, Thermo Scientific Chemicals

Catalog number: A11123.22
100 g, Each
Thermo Scientific Chemicals

Antimony(III) oxide, 99%, Thermo Scientific Chemicals

Catalog number: A11123.22
100 g, Each
Quantity
Catalog number: A11123.22
also known as A11123-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1309-64-4
IUPAC Name
diantimony(3+) trioxidandiide
Molecular Formula
O3Sb2
InChI Key
GHPGOEFPKIHBNM-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Sb+3].[Sb+3]
Appearance (Color)
White
Form
Powder
Assay from Supplier's CofA
≥98.5 to ≤101.5%

Description

Antimony(III) oxide is used as a catalyst, vulcanization of rubber, an opacifying agent for glasses, ceramics and enamels. It is used as a flame retardant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Antimony(III) oxide is used as a catalyst, vulcanization of rubber, an opacifying agent for glasses, ceramics and enamels. It is used as a flame retardant.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. Keep container tightly closed.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text