Glycinamide hydrochloride is a buffer that is useful in the physiological pH range. And it is also used as a pharmaceutical intermediate, in the organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Glycinamide hydrochloride is a buffer that is useful in the physiological pH range. And it is also used as a pharmaceutical intermediate, in the organic synthesis.
Solubility
Soluble in water (1100g/L).
Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Michael Granvogl.; Peter Schieberle. Thermally Generated 3-Aminopropionamide as a Transient Intermediate in the Formation of Acrylamide. J. Agric. Food Chem. 2006, 54, (16), 5933-5938.
Norman E. Good.; G. Douglas Winget.; Wilhelmina Winter.; Thomas N. Connolly.; Seikichi Izawa.; Raizada M. M. Singh. Hydrogen Ion Buffers for Biological Research. Biochemistry. 1966, 5, (2), 467-477.