Thermo Scientific Chemicals

Glycinamide hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A11128.22
100 g, Each
Thermo Scientific Chemicals

Glycinamide hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A11128.22
100 g, Each
Quantity
Catalog number: A11128.22
also known as A11128-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1668-10-6
IUPAC Name
hydrogen 2-aminoacetamide chloride
Molecular Formula
C2H7ClN2O
InChI Key
WKNMKGVLOWGGOU-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NCC(N)=O
Appearance (Color)
White to pale cream
Clarity
0.5% w/v solution in water is transparent to almost transparent.
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤1.0%
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%

Description

Glycinamide hydrochloride is a buffer that is useful in the physiological pH range. And it is also used as a pharmaceutical intermediate, in the organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycinamide hydrochloride is a buffer that is useful in the physiological pH range. And it is also used as a pharmaceutical intermediate, in the organic synthesis.

Solubility
Soluble in water (1100g/L).

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text