Thermo Scientific Chemicals

tert-Butylamine, 98%, Thermo Scientific Chemicals

Catalog number: A11135.AU
1000 mL, Each
Thermo Scientific Chemicals

tert-Butylamine, 98%, Thermo Scientific Chemicals

Catalog number: A11135.AU
1000 mL, Each
Quantity
Catalog number: A11135.AU
also known as A11135-AU
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-64-9
IUPAC Name
2-methylpropan-2-amine
Molecular Formula
C4H11N
InChI Key
YBRBMKDOPFTVDT-UHFFFAOYSA-N
SMILES
CC(C)(C)N
Appearance (Color)
Clear colorless
Refractive Index
1.3760-1.3805 @ 20?C
Assay (GC)
≥97.5%
Form
Liquid
Identification (FTIR)
Conforms

Description

tert-butylamine is used as a precursor for the preparation of rubber accelerators, pharmaceuticals and dyes. It is employed in synthesis of N-tert-butyl-acetamide, isoquinolines, pyridines, decumbenine, sulfontes, sulfonamides and methyl beta-ketophosphonate. It is also used in the study the effect of amines in influencing atmospheric sulfuric acid - water nucleation, hydroamination of terminal alkynes and measuring the surface acidity of solid catalyst. It acts as an indicator molecule in the study of NMR -based pH determination method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-butylamine is used as a precursor for the preparation of rubber accelerators, pharmaceuticals and dyes. It is employed in synthesis of N-tert-butyl-acetamide, isoquinolines, pyridines, decumbenine, sulfontes, sulfonamides and methyl beta-ketophosphonate. It is also used in the study the effect of amines in influencing atmospheric sulfuric acid - water nucleation, hydroamination of terminal alkynes and measuring the surface acidity of solid catalyst. It acts as an indicator molecule in the study of NMR -based pH determination method.

Solubility
Miscible with water, alcohol, ether, chloroform and organic solvents.

Notes
Air sensitive. Incompatible with strong oxidizing agents, strong acids and acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text