Thermo Scientific Chemicals

Dopamine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A11136.22
100 g, Each
Thermo Scientific Chemicals

Dopamine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A11136.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A11136.22
also known as A11136-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
62-31-7
IUPAC Name
hydrogen 4-(2-aminoethyl)benzene-1,2-diol chloride
Molecular Formula
C8H12ClNO2
InChI Key
CTENFNNZBMHDDG-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NCCC1=CC=C(O)C(O)=C1
Loss on Drying
≤1% (105?C/constant weight)
Assay (Titration ex Chloride)
≥98.5 to ≤101.5%
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to pale cream or pale pink

Description

A neurotransmitter and vasopressor that modulates cortical activation. Dopamine hydrochloride is a vasopressor that moderates cortical activation. It is used as a precursor to norepinepherine and epinephrine. It is an important neurotransmitter and chemical messenger that helps in the transmission of signals in the brain and other vital areas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A neurotransmitter and vasopressor that modulates cortical activation. Dopamine hydrochloride is a vasopressor that moderates cortical activation. It is used as a precursor to norepinepherine and epinephrine. It is an important neurotransmitter and chemical messenger that helps in the transmission of signals in the brain and other vital areas.

Solubility
Soluble in water, methanol and dimethyl sulfoxide. Insoluble in ether, petroleum ether, chloroform, benzene and toluene.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text