3-Iodobenzyl bromide, 96%, Thermo Scientific Chemicals
3-Iodobenzyl bromide, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Iodobenzyl bromide, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A11140.06
also known as A11140-06
Price (USD)/ Each
89.65
Online exclusive
99.40 
Save 9.75 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
89.65
Online exclusive
99.40 
Save 9.75 (10%)
Add to cart
3-Iodobenzyl bromide, 96%, Thermo Scientific Chemicals
Catalog numberA11140.06
Price (USD)/ Each
89.65
Online exclusive
99.40 
Save 9.75 (10%)
-
Add to cart
Chemical Identifiers
CAS49617-83-6
IUPAC Name1-(bromomethyl)-3-iodobenzene
Molecular FormulaC7H6BrI
InChI KeyBACZSVQZBSCWIG-UHFFFAOYSA-N
SMILESBrCC1=CC(I)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow to pale brown
Assay (GC)≥95.0%
FormCrystalline powder or powder
Melting Point (clear melt)48-52°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only