Carbohydrazide, 97%, Thermo Scientific Chemicals
Carbohydrazide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Carbohydrazide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number A11145.22
also known as A11145-22
Price (USD)/ Each
133.65
Online exclusive
148.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
133.65
Online exclusive
148.00 
Save 14.35 (10%)
Add to cart
Carbohydrazide, 97%, Thermo Scientific Chemicals
Catalog numberA11145.22
Price (USD)/ Each
133.65
Online exclusive
148.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
CAS497-18-7
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)White
Melting Point (clear melt)150.0-160.0?C
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
View more
Carbohydrazide is widely utilized as a curing agent for epoxide-type resins and as an oxygen scrubber in boiler systems. It is applied in photography to stabilize color developers that produce images of the azo-methine and azine classes. It is used to develop ammunition propellants and stabilize soaps. It is also a useful reagent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carbohydrazide is widely utilized as a curing agent for epoxide-type resins and as an oxygen scrubber in boiler systems. It is applied in photography to stabilize color developers that produce images of the azo-methine and azine classes. It is used to develop ammunition propellants and stabilize soaps. It is also a useful reagent in organic synthesis.

Solubility
Soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents, strong acids, copper, zinc, nickel, lead and brass.
RUO – Research Use Only

General References:

  1. Li, Z.; Huang, H.; Zhang, T.; Zhang, G.; Zhang, F. Electric-field-induced structural and electronic changes and decomposition of an energetic complex: a computational study on zinc carbohydrazide perchlorate crystals. RSC Adv. 2015, 5 (29), 22601-22608.
  2. Qi, J.; Benipal, N.; Chadderdon, D. J.; Huo, J.; Jiang, Y.; Qiu, Y.; Han, X.; Hu, Y. H.; Shanks, B. H.; Li, W. Carbon nanotubes as catalysts for direct carbohydrazide fuel cells. Carbon 2015, 89, 142-147.