Thermo Scientific Chemicals

1-Octene, 97+%, Thermo Scientific Chemicals

Catalog number: A11146.0F
2500 mL, Each
Thermo Scientific Chemicals

1-Octene, 97+%, Thermo Scientific Chemicals

Catalog number: A11146.0F
2500 mL, Each
Quantity
Catalog number: A11146.0F
also known as A11146-0F
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-66-0
IUPAC Name
oct-1-ene
Molecular Formula
C8H16
InChI Key
KWKAKUADMBZCLK-UHFFFAOYSA-N
SMILES
CCCCCCC=C
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.4060-1.4110 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥97.0%

Description

1-Octene acts as a comonomer used in the preparation of polyethylene, especially high-density polyethylene (HDPE) and linear low-density polyethylene(LLDPE) resins. It is also used in organic synthesis, surfactants and plasticizers. Further, it is used in process regulators and viscosity adjustors. In addition to this, it is used in the preparation of linear aldehyde through the oxo synthesis (hydroformylation) to get the nonanal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octene acts as a comonomer used in the preparation of polyethylene, especially high-density polyethylene (HDPE) and linear low-density polyethylene(LLDPE) resins. It is also used in organic synthesis, surfactants and plasticizers. Further, it is used in process regulators and viscosity adjustors. In addition to this, it is used in the preparation of linear aldehyde through the oxo synthesis (hydroformylation) to get the nonanal.

Solubility
Miscible with water, ether, alcohol and acetone.

Notes
Incompatible with bromine, chlorine, acids and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text