2-Butenoic acid, 4-oxo-4-phenyl- could react with morpholine, and obtain the 2-morpholin-4-yl-4-oxo-4-phenyl-butyric acid. This reaction needs the solvent of toluene, ethanol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Butenoic acid, 4-oxo-4-phenyl- could react with morpholine, and obtain the 2-morpholin-4-yl-4-oxo-4-phenyl-butyric acid. This reaction needs the solvent of toluene, ethanol.
Solubility
Insoluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only
George A. Olah. Stable Carbonium Ions. IX.1 Methylbenzenonium Hexafluoroantimonates2. J. Am. Chem. Soc. 1965, 87, (5), 1103-1108.
Leonard Steinbach.; Ernest I. Becker. A Synthesis for β-Aroylacrylic Acids Substituted with Electron-withdrawing Groups. J. Am. Chem. Soc. 1954, 76, (22), 5808-5810.