Thermo Scientific Chemicals

2,5-Dichloroaniline, 99%, Thermo Scientific Chemicals

Catalog number: A11151.36
500 g, Each
Thermo Scientific Chemicals

2,5-Dichloroaniline, 99%, Thermo Scientific Chemicals

Catalog number: A11151.36
500 g, Each
Quantity
Catalog number: A11151.36
also known as A11151-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
95-82-9
Assay (GC)
≥98.5%
Form
Flakes or fused solid
Appearance (Color)
White to brown to grey to black
Melting Point (clear melt)
46.0-52.0?C

Description

2,5-Dichloroaniline was used in a study to develop a fast and sensitive quantitative method for the detection of some aniline derivatives by solid-phase microextraction in gas chromatography-mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dichloroaniline was used in a study to develop a fast and sensitive quantitative method for the detection of some aniline derivatives by solid-phase microextraction in gas chromatography-mass spectrometry.

Solubility
Soluble in dilute hydrochloric acid.

Notes
Air & Light Sensitive, keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text