Thermo Scientific Chemicals

Glutaric anhydride, 98%, Thermo Scientific Chemicals

Catalog number: A11152.0B
1000 g, Each
Thermo Scientific Chemicals

Glutaric anhydride, 98%, Thermo Scientific Chemicals

Catalog number: A11152.0B
1000 g, Each
Quantity
Catalog number: A11152.0B
also known as A11152-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-55-4
IUPAC Name
oxane-2,6-dione
Molecular Formula
C5H6O3
InChI Key
VANNPISTIUFMLH-UHFFFAOYSA-N
SMILES
O=C1CCCC(=O)O1
Form
Crystals or crystalline powder or powder
Appearance (Color)
White to pale yellow
Assay (GC)
≥97.5% (non-U.S. specification)
Assay from Suppliers CofA
≥97.5% (U.S. specification)
Comment
Purchased in the U.S. and in other countries

Description

Glutaric anhydride is used in the production of glutaric acid family pharmaceuticals. Its derivatives are also used in certain corrosion inhibitors, some surfactants and metal finishing compounds. It is also used in the production of polymers, particularly modified unsaturated linear (epoxy) polyesters and as a raw material in the manufacture of flavors. It is also used in improvement of electrode/electrolyte interfaces in High-Voltage Spinel Lithium-Ion batteries as an electrolyte additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glutaric anhydride is used in the production of glutaric acid family pharmaceuticals. Its derivatives are also used in certain corrosion inhibitors, some surfactants and metal finishing compounds. It is also used in the production of polymers, particularly modified unsaturated linear (epoxy) polyesters and as a raw material in the manufacture of flavors. It is also used in improvement of electrode/electrolyte interfaces in High-Voltage Spinel Lithium-Ion batteries as an electrolyte additive.

Solubility
Soluble in Ether, dimethyl sulfoxide and methanol and water.

Notes
Moisture sensitive. Incompatible withstrong oxidizing agents, acids, bases, alcohols and amines.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text