Thermo Scientific Chemicals

2-Amino-5-chlorobenzamide, 98+%, Thermo Scientific Chemicals

Catalog number: A11153.03
1 g, Each
Thermo Scientific Chemicals

2-Amino-5-chlorobenzamide, 98+%, Thermo Scientific Chemicals

Catalog number: A11153.03
1 g, Each
Quantity
Catalog number: A11153.03
also known as A11153-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5202-85-7
IUPAC Name
2-amino-5-chlorobenzamide
Molecular Formula
C7H7ClN2O
InChI Key
DNRVZOZGQHHDAT-UHFFFAOYSA-N
SMILES
NC(=O)C1=CC(Cl)=CC=C1N
Appearance (Color)
White to pale yellow to yellow to brown to gray
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0%
Melting Point (clear melt)
168.0-175.0?C
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥98.0%

Description

2-Amino-5-chlorobenzamide react with 2-Azido-benzoyl chloride to get 2-Azido-5'-chloro-2'-carboxamidobenzanilide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-chlorobenzamide react with 2-Azido-benzoyl chloride to get 2-Azido-5′-chloro-2′-carboxamidobenzanilide.

Solubility
Soluble in acetone.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text