Thermo Scientific Chemicals

Molybdenum(VI) oxide, 99.5%, Thermo Scientific Chemicals

Catalog number: A11159.30
250 g, Each
Thermo Scientific Chemicals

Molybdenum(VI) oxide, 99.5%, Thermo Scientific Chemicals

Catalog number: A11159.30
250 g, Each
Quantity
Catalog number: A11159.30
also known as A11159-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1313-27-5
IUPAC Name
trioxomolybdenum
Molecular Formula
MoO3
InChI Key
JKQOBWVOAYFWKG-UHFFFAOYSA-N
SMILES
O=[Mo](=O)=O
Form
Powder
Appearance (Color)
White to grey
Assay from Suppliers CofA
≥99.5%

Description

Molybdenum(VI) oxide is a very promising and stable oxide for electrochromic applications and used as a catalyst in the oil industry. It is also used to make metal molybdenum. It has a wide band gap type semiconductor with distinctive electrochromic, thermochromic, and photochromic properties, has been widely investigated as smart materials for catalysis sensors, lubricants, lithium battery, organic solar cells, and display materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Molybdenum(VI) oxide is a very promising and stable oxide for electrochromic applications and used as a catalyst in the oil industry. It is also used to make metal molybdenum. It has a wide band gap type semiconductor with distinctive electrochromic, thermochromic, and photochromic properties, has been widely investigated as smart materials for catalysis sensors, lubricants, lithium battery, organic solar cells, and display materials.

Solubility
Slightly soluble in water. Soluble in concentrated mineral acids and alkali hydroxide solutions.

Notes
It does not react with acids other than hydrofluoric and concentrated sulfuric acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text