4'-Methoxyacetophenone is used as a component of perfumes and as chemical intermediate in the manufacture of pharmaceuticals, resins, flavouring agents. And also used to study ruthenium catalyzed step growth copolymerization of 4?-methoxyacetophenone with α,ω-dienes to yield copolymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4′-Methoxyacetophenone is used as a component of perfumes and as chemical intermediate in the manufacture of pharmaceuticals, resins, flavouring agents. And also used to study ruthenium catalyzed step growth copolymerization of 4′-methoxyacetophenone with α,ω-dienes to yield copolymers.
Solubility
Insoluble in water.
Notes
Light Sensitive, Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
- Wei Wang.; Min-Hua Zong.; Wen-Yong Lou. Use of an ionic liquid to improve asymmetric reduction of 4'-methoxyacetophenone catalyzed by immobilized Rhodotorulasp. AS2.2241 cells. Journal of Molecular Catalysis B: Enzymatic. 2009, 56, (1), 70-76.
- Godman, J.L.; Horton, D.; Tronchet, J.M.J. Extension of sugar chains through acetylenic intermediates ☆: III. 1-heptyne-L-gluco- and L-manno-3,4,5,6,7-pentol from 2,3,4,5-tetra-O-benzoyl-aldehydo-L-arabinose. Carbohydrate Research. 1967, 4, (5), 392-400.