2'-Deoxyadenosine monohydrate, 99%
2'-Deoxyadenosine monohydrate, 99%
Thermo Scientific Chemicals

2'-Deoxyadenosine monohydrate, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A11166.03
also known as A11166-03
Price (USD)/ Each
35.65
Online exclusive
39.00 
Save 3.35 (9%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
35.65
Online exclusive
39.00 
Save 3.35 (9%)
Add to cart
2'-Deoxyadenosine monohydrate, 99%
Catalog numberA11166.03
Price (USD)/ Each
35.65
Online exclusive
39.00 
Save 3.35 (9%)
-
Add to cart
Chemical Identifiers
CAS16373-93-6
IUPAC Name(2R,3S,5R)-5-(6-amino-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Molecular FormulaC10H13N5O3
InChI KeyOLXZPDWKRNYJJZ-YUZWJPFSNA-N
SMILESNC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)5.0 to 8.0% (theory 6.69%)
Optical Rotation-25.3? ± 1.5? (c = 0.5, Water)
Appearance (Color)White
Assay (HPLC)≥98.5%
Melting Point (clear melt)188.0-194.0?C
View more
2?-Deoxyadenosineise is used by some cells as an energy source under energy stress conditions and to affect cAMP levels. 2?-dAdo is used in comparison studies of the functions of adenosine analogues on various biological processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Deoxyadenosineise is used by some cells as an energy source under energy stress conditions and to affect cAMP levels. 2′-dAdo is used in comparison studies of the functions of adenosine analogues on various biological processes.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Ashley M. Berman.; Jeffrey S. Johnson. Copper-Catalyzed Electrophilic Amination of Organozinc Nucleophiles: Documentation of O-Benzoyl Hydroxylamines as Broadly Useful R2N(+) and RHN(+) Synthons. J. Org. Chem. 2006, 71, (1), 219-224.
  2. Songbai Liu, Lanny S. Liebeskind. A Simple, Modular Synthesis of Substituted Pyridines. J. Am. Chem. Soc. 2008, 130, (22), 6918-6919.