Thermo Scientific Chemicals

Vanillin, 99%, Thermo Scientific Chemicals

Catalog number: A11169.0E
2500 g, Each
Thermo Scientific Chemicals

Vanillin, 99%, Thermo Scientific Chemicals

Catalog number: A11169.0E
2500 g, Each
Quantity
Catalog number: A11169.0E
also known as A11169-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
121-33-5
IUPAC Name
4-hydroxy-3-methoxybenzaldehyde
Molecular Formula
C8H8O3
InChI Key
MWOOGOJBHIARFG-UHFFFAOYSA-N
SMILES
COC1=CC(C=O)=CC=C1O
Comment
Material sourced in the U.S. and in other countries
Form
Crystals or crystalline powder
Appearance (Color)
White to cream or pale yellow
Assay (GC)
≥98.5% (non-U.S. sourced material)
Comment
Material sourced in the U.S. and in other countries

Description

Vanillin is a flavoring agent used in sweet foods, ice cream and chocolate. It is used in perfumes as well as to control the unpleasant odors in medicines. It acts as a chemical intermediate in the preparation of pharmaceuticals. In thin layer chromatography, it is used as a general-purpose stain for developing TLC plates to enable in visualizing components of a reaction mixture. Similarly, vanillin hydro chloride staining can be used to visualize the localization of tannins in cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vanillin is a flavoring agent used in sweet foods, ice cream and chocolate. It is used in perfumes as well as to control the unpleasant odors in medicines. It acts as a chemical intermediate in the preparation of pharmaceuticals. In thin layer chromatography, it is used as a general-purpose stain for developing TLC plates to enable in visualizing components of a reaction mixture. Similarly, vanillin hydro chloride staining can be used to visualize the localization of tannins in cells.

Solubility
Soluble in water, diethyl ether, acetone, methanol, benzene, petroleum ether, chloroform, carbon disulfide, glacial acetic acid, pyridine and in aqueous solutions of alkali hydroxides.

Notes
Moisture and light sensitive. Incompatible with oxidizing agents, alkalis and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text