Thermo Scientific Chemicals

1-Decyne, 98%, Thermo Scientific Chemicals

Catalog number: A11171.14
25 g, Each
Thermo Scientific Chemicals

1-Decyne, 98%, Thermo Scientific Chemicals

Catalog number: A11171.14
25 g, Each
Quantity
Catalog number: A11171.14
also known as A11171-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
764-93-2
IUPAC Name
dec-1-yne
Molecular Formula
C10H18
InChI Key
ILLHQJIJCRNRCJ-UHFFFAOYSA-N
SMILES
CCCCCCCCC#C
Form
Liquid
Refractive Index
1.4250-1.4290 @ 20?C
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow

Description

1-Decyne has been used to stabilize ruthenium nanoparticles by forming ruthenium-vinylidene interfacial bonds. It is also used as primary and secondary intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Decyne has been used to stabilize ruthenium nanoparticles by forming ruthenium-vinylidene interfacial bonds. It is also used as primary and secondary intermediates.

Solubility
Not miscible in water.

Notes
Incompatible with strong oxidizing agents. Store at 0-6°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text