Thermo Scientific Chemicals

Benzonitrile, 99%, Thermo Scientific Chemicals

Catalog number: A11173.36
500 g, Each
Thermo Scientific Chemicals

Benzonitrile, 99%, Thermo Scientific Chemicals

Catalog number: A11173.36
500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A11173.36
also known as A11173-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-47-0
IUPAC Name
benzonitrile
Molecular Formula
C7H5N
InChI Key
JFDZBHWFFUWGJE-UHFFFAOYSA-N
SMILES
N#CC1=CC=CC=C1
Assay from Supplier's CofA
≥98.5% (US-Sourced Material)
Refractive Index
1.5275-1.5305 @ 20?C (UK-Sourced Material)
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5% (UK-Sourced Material)
Comment
Material Sourced in UK and US

Description

Benzonitrile is a widely utilized as a solvent and an intermediate in industries making drugs, perfumes, dyes, rubber, textiles, resins and specialty lacquers. It finds application as a versatile precursor for many derivatives. It coordinates with transition metal to form complexes which act as synthetic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzonitrile is a widely utilized as a solvent and an intermediate in industries making drugs, perfumes, dyes, rubber, textiles, resins and specialty lacquers. It finds application as a versatile precursor for many derivatives. It coordinates with transition metal to form complexes which act as synthetic intermediates.

Solubility
Miscible with common organic solvents. Immiscible with water.

Notes
Hygroscopic. Incompatible with strong bases, strong oxidizing agents, reducing agents, acids, chlorates, nitrates and plastics.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text