Thermo Scientific Chemicals

2,6-Dichloropyrazine, 98%, Thermo Scientific Chemicals

Catalog number: A11175.14
25 g, Each
Thermo Scientific Chemicals

2,6-Dichloropyrazine, 98%, Thermo Scientific Chemicals

Catalog number: A11175.14
25 g, Each
Quantity
Catalog number: A11175.14
also known as A11175-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4774-14-5
IUPAC Name
2,6-dichloropyrazine
Molecular Formula
C4H2Cl2N2
InChI Key
LSEAAPGIZCDEEH-UHFFFAOYSA-N
SMILES
ClC1=CN=CC(Cl)=N1
Appearance (Color)
White to cream to pale yellow to pale brown
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
51-56?C

Description

2,6-Dichloropyrazine is used in the preparation of pyrazine and pyridine compounds as ATR kinase inhibitors. It is also used in the synthesis of oxacalixarenes containing nitrogen heterocycles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dichloropyrazine is used in the preparation of pyrazine and pyridine compounds as ATR kinase inhibitors. It is also used in the synthesis of oxacalixarenes containing nitrogen heterocycles.

Solubility
Soluble in ethanol and methanol.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong acids and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text