Thermo Scientific Chemicals

1-Hexadecanol, 98%, Thermo Scientific Chemicals

Catalog number: A11180.36
500 g, Each
Thermo Scientific Chemicals

1-Hexadecanol, 98%, Thermo Scientific Chemicals

Catalog number: A11180.36
500 g, Each
Quantity
Catalog number: A11180.36
also known as A11180-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
36653-82-4
IUPAC Name
hexadecan-1-ol
Molecular Formula
C16H34O
InChI Key
BXWNKGSJHAJOGX-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCO
Appearance (Color)
White
Assay (GC)
≥97.5%
Melting Point (clear melt)
47.0-53.0?C
Form
Flakes or pellets
Identification (FTIR)
Conforms

Description

1-Hexadecanol is used as cosmetic ingredient, emollient, foam stabilizer. It is also used as the raw material for the manufacture of surfactants. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexadecanol is used as cosmetic ingredient, emollient, foam stabilizer. It is also used as the raw material for the manufacture of surfactants. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Soluble in alcohol, chloroform, ether

Notes
Keep away from direct sunlight. Incompatible with strong oxidizing agents, strong acids. Store at 2-8°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text