Thermo Scientific Chemicals

(+/-)-1,1'-Bi(2-naphthol), 99%, Thermo Scientific Chemicals

Catalog number: A11182.22
100 g, Each
Thermo Scientific Chemicals

(+/-)-1,1'-Bi(2-naphthol), 99%, Thermo Scientific Chemicals

Catalog number: A11182.22
100 g, Each
Quantity
Catalog number: A11182.22
also known as A11182-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
602-09-5
IUPAC Name
[1,1'-binaphthalene]-2,2'-diol
Molecular Formula
C20H14O2
InChI Key
PPTXVXKCQZKFBN-UHFFFAOYSA-N
SMILES
OC1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1O
Form
Powder
Melting Point (clear melt)
214.0-221.0°C
Appearance (Color)
White to pale cream to pale brown
Assay (HPLC)
≥98.5%

Description

(±)-1,1'-Bi(2-naphthol) acts as chiral separation agents. It is widely used in chiral liquid crystal,medicines,spices etc,.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-1,1′-Bi(2-naphthol) acts as chiral separation agents. It is widely used in chiral liquid crystal,medicines,spices etc,.

Solubility
Soluble in dioxane (50 mg/mL). Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Refrigerated (2-8°C)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text