4-Fluorobenzophenone, 97%, Thermo Scientific Chemicals
4-Fluorobenzophenone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Fluorobenzophenone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number A11185.22
also known as A11185-22
Price (USD)/ Each
148.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
148.00
Add to cart
4-Fluorobenzophenone, 97%, Thermo Scientific Chemicals
Catalog numberA11185.22
Price (USD)/ Each
148.00
-
Add to cart
Chemical Identifiers
CAS345-83-5
IUPAC Name(4-fluorophenyl)(phenyl)methanone
Molecular FormulaC13H9FO
InChI KeyOGTSHGYHILFRHD-UHFFFAOYSA-N
SMILESFC1=CC=C(C=C1)C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)White to cream
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)43-49?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only