Thermo Scientific Chemicals

Ethyl dodecanoate, 98%, Thermo Scientific Chemicals

Catalog number: A11188.0F
2500 mL, Each
Thermo Scientific Chemicals

Ethyl dodecanoate, 98%, Thermo Scientific Chemicals

Catalog number: A11188.0F
2500 mL, Each
Quantity
Catalog number: A11188.0F
also known as A11188-0F
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
106-33-2
IUPAC Name
ethyl dodecanoate
Molecular Formula
C14H28O2
InChI Key
MMXKVMNBHPAILY-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCC(=O)OCC
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Refractive Index
1.4290-1.4330 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text